Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACCAAGTGCTGCTGGGCCGGAGC[A/G]GCCCCTCCAAGAACTACGTGATGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601597 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTG2 - BTG anti-proliferation factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006763.2 | 510 | Missense Mutation | AGC,GGC | S,G 147 | NP_006754.1 |
LINC01136 - long intergenic non-protein coding RNA 1136 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |