Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGAGGTATGTTGAGGGGAAAAG[A/C]AGCAGCAATTTTAAAGCAATCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615254 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBED9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZBED9 - zinc finger BED-type containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052923.1 | 3821 | Silent Mutation | CTG,CTT | L,L 1269 | NP_443155.1 | |
XM_011514283.2 | 3821 | Silent Mutation | CTG,CTT | L,L 1118 | XP_011512585.1 | |
XM_011514284.2 | 3821 | Silent Mutation | CTG,CTT | L,L 1118 | XP_011512586.1 | |
XM_011514285.2 | 3821 | Silent Mutation | CTG,CTT | L,L 1118 | XP_011512587.1 | |
XM_011514286.2 | 3821 | Silent Mutation | CTG,CTT | L,L 1118 | XP_011512588.1 | |
XM_011514287.2 | 3821 | Silent Mutation | CTG,CTT | L,L 1118 | XP_011512589.1 | |
XM_011514288.2 | 3821 | Silent Mutation | CTG,CTT | L,L 1073 | XP_011512590.1 | |
XM_017010231.1 | 3821 | Silent Mutation | CTG,CTT | L,L 1118 | XP_016865720.1 |