Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCGGAACTCCACGAGTGACCTCG[C/G]GATCTGGCCCGGCTCCCGCTCGTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610493 MIM: 608770 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIXDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DIXDC1 - DIX domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DLAT - dihydrolipoamide S-acetyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001931.4 | 768 | Missense Mutation | CGA,GGA | R,G 37 | NP_001922.2 | |
XM_011542647.2 | 768 | Missense Mutation | CGA,GGA | R,G 37 | XP_011540949.1 |