Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATATTTGCTTGATATGCTCCAAT[A/C]GGAATTTCAGGCAATCCAATGTAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GIN1 - gypsy retrotransposon integrase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317954.1 | 1250 | Silent Mutation | NP_001304883.1 | |||
NM_017676.2 | 1250 | Silent Mutation | NP_060146.2 | |||
XM_005272028.3 | 1250 | Silent Mutation | XP_005272085.1 | |||
XM_017009599.1 | 1250 | Silent Mutation | XP_016865088.1 |