Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTATCACCCAGGGGCTTCAATA[C/T]CTGTGGTATGAGAAATGTGCAAAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609368 | ||||||||||||||||||||
Literature Links: |
ATL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATL2 - atlastin GTPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308076.1 | 2565 | Missense Mutation | ATA,GTA | I,V 527 | NP_001295005.1 | |
XM_011533017.1 | 2565 | UTR 3 | XP_011531319.1 | |||
XM_011533018.2 | 2565 | UTR 3 | XP_011531320.1 | |||
XM_011533019.2 | 2565 | UTR 3 | XP_011531321.1 | |||
XM_011533020.1 | 2565 | UTR 3 | XP_011531322.1 | |||
XM_011533022.2 | 2565 | UTR 3 | XP_011531324.1 | |||
XM_011533023.2 | 2565 | UTR 3 | XP_011531325.1 | |||
XM_017004686.1 | 2565 | UTR 3 | XP_016860175.1 | |||
XM_017004687.1 | 2565 | UTR 3 | XP_016860176.1 | |||
XM_017004688.1 | 2565 | Missense Mutation | ATA,GTA | I,V 318 | XP_016860177.1 |