Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAACTCAGATTCTTCCTCAGCTAG[G/T]GTAGAATCCTTGGAACAGTGGAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615295 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHSA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AHSA2 - AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321300.1 | 2553 | Intron | NP_001308229.1 | |||
NM_001321301.1 | 2553 | Intron | NP_001308230.1 | |||
NM_001321302.1 | 2553 | Intron | NP_001308231.1 | |||
NM_001321303.1 | 2553 | Intron | NP_001308232.1 | |||
NM_001321304.1 | 2553 | Intron | NP_001308233.1 | |||
NM_001321305.1 | 2553 | UTR 3 | NP_001308234.1 | |||
NM_152392.4 | 2553 | Intron | NP_689605.1 | |||
XM_011532541.2 | 2553 | UTR 3 | XP_011530843.1 | |||
XM_011532543.2 | 2553 | UTR 3 | XP_011530845.1 | |||
XM_011532545.2 | 2553 | UTR 3 | XP_011530847.1 | |||
XM_017003377.1 | 2553 | UTR 3 | XP_016858866.1 | |||
XM_017003378.1 | 2553 | UTR 3 | XP_016858867.1 |
USP34 - ubiquitin specific peptidase 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014709.3 | 2553 | Silent Mutation | ACA,ACC | T,T 3460 | NP_055524.3 |