Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180630 MIM: 604983 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP95 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP95 - centrosomal protein 95 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DDX5 - DEAD-box helicase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320595.1 | 1299 | Silent Mutation | CAC,CAT | H,H 299 | NP_001307524.1 | |
NM_001320596.1 | 1299 | Silent Mutation | CAC,CAT | H,H 299 | NP_001307525.1 | |
NM_001320597.1 | 1299 | Silent Mutation | CAC,CAT | H,H 299 | NP_001307526.1 | |
NM_004396.4 | 1299 | Silent Mutation | CAC,CAT | H,H 299 | NP_004387.1 |
MIR3064 - microRNA 3064 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5047 - microRNA 5047 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLG2 - DNA polymerase gamma 2, accessory subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |