Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATGAATATACTCTGGAAATTAAT[A/G]GGAAAAGTCTCAAGAAGTATATGGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
FAIM PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FAIM - Fas apoptotic inhibitory molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033030.1 | 941 | Missense Mutation | AGG,GGG | R,G 117 | NP_001028202.1 | |
NM_001033031.1 | 941 | Missense Mutation | AGG,GGG | R,G 105 | NP_001028203.1 | |
NM_001033032.1 | 941 | Missense Mutation | AGG,GGG | R,G 83 | NP_001028204.1 | |
NM_018147.3 | 941 | Missense Mutation | AGG,GGG | R,G 83 | NP_060617.1 | |
XM_011512950.2 | 941 | Missense Mutation | AGG,GGG | R,G 137 | XP_011511252.1 |