Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCACAAAGTTTTTATGTGGATCA[A/G]TATCCTCGGAAAGAATTAGACTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612424 MIM: 607789 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EYS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EYS - eyes shut homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142800.1 | 9810 | Missense Mutation | ACT,ATT | T,I 3070 | NP_001136272.1 | |
NM_001142801.1 | 9810 | Intron | NP_001136273.1 | |||
NM_001292009.1 | 9810 | Missense Mutation | ACT,ATT | T,I 3091 | NP_001278938.1 | |
NM_198283.1 | 9810 | Intron | NP_938024.1 |
PHF3 - PHD finger protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |