Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCCCCTGGCTCCCGTCTCTCAC[C/T]TCTGGGTCCCCATCCTCAGGGGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SBK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SBK2 - SH3 domain binding kinase family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SBK3 - SH3 domain binding kinase family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199824.2 | 45 | Silent Mutation | GAA,GAG | E,E 15 | NP_001186753.1 | |
XM_011526298.2 | 45 | Intron | XP_011524600.1 |