Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCAGCTGAGCCGAAAGAGTGTG[A/G]ATGGCTTCCTGGTCTGGGTACAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608148 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SATB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SATB2 - SATB homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172509.1 | 2348 | Silent Mutation | ATC,ATT | I,I 645 | NP_001165980.1 | |
NM_001172517.1 | 2348 | Silent Mutation | ATC,ATT | I,I 645 | NP_001165988.1 | |
NM_015265.3 | 2348 | Silent Mutation | ATC,ATT | I,I 645 | NP_056080.1 | |
XM_005246396.2 | 2348 | Silent Mutation | ATC,ATT | I,I 587 | XP_005246453.1 | |
XM_006712372.2 | 2348 | Silent Mutation | ATC,ATT | I,I 645 | XP_006712435.1 | |
XM_011510840.2 | 2348 | Silent Mutation | ATC,ATT | I,I 645 | XP_011509142.1 | |
XM_017003656.1 | 2348 | Silent Mutation | ATC,ATT | I,I 587 | XP_016859145.1 |