Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAAGATGTGCTATGGCTGTGTC[A/G]AAAACAAGGCAGTCGCTGTTCATGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602213 | |||||||||||||||||||||||
Literature Links: |
LOC101928105 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LOC101928105 - uncharacterized LOC101928105 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIAH2 - siah E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005067.5 | 1524 | Silent Mutation | TTC,TTT | F,F 299 | NP_005058.3 |