Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGGAGCCCGAAGAGCCACACTC[C/T]GAGGGGGCATCGCAGGAGGATGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603436 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF205 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF205 - zinc finger protein 205 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042428.1 | 344 | Silent Mutation | TCC,TCT | S,S 58 | NP_001035893.1 | |
NM_001278158.1 | 344 | Silent Mutation | TCC,TCT | S,S 58 | NP_001265087.1 | |
NM_003456.2 | 344 | Silent Mutation | TCC,TCT | S,S 58 | NP_003447.2 | |
XM_005255558.2 | 344 | Silent Mutation | TCC,TCT | S,S 58 | XP_005255615.1 |
ZNF205-AS1 - ZNF205 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |