Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGTAGGCGGCGGCGGCGGCACAG[A/G]CACAGCCCCACCGGCCCGCCAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300080 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBM10 - RNA binding motif protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204466.1 | 423 | Intron | NP_001191395.1 | |||
NM_001204467.1 | 423 | Silent Mutation | AGA,AGG | R,R 87 | NP_001191396.1 | |
NM_001204468.1 | 423 | Silent Mutation | AGA,AGG | R,R 152 | NP_001191397.1 | |
NM_005676.4 | 423 | Silent Mutation | AGA,AGG | R,R 87 | NP_005667.2 | |
NM_152856.2 | 423 | Intron | NP_690595.1 | |||
XM_005272677.4 | 423 | Silent Mutation | AGA,AGG | R,R 152 | XP_005272734.1 | |
XM_005272678.4 | 423 | Intron | XP_005272735.1 | |||
XM_005272679.4 | 423 | Intron | XP_005272736.1 | |||
XM_006724563.1 | 423 | Intron | XP_006724626.1 | |||
XM_011543989.2 | 423 | Intron | XP_011542291.1 | |||
XM_017029884.1 | 423 | UTR 5 | XP_016885373.1 | |||
XM_017029885.1 | 423 | UTR 5 | XP_016885374.1 |