Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGACCTGCGCGCCGTGCACCAGCA[C/G]CTGTGCTCGGTGAACTCGCAGTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604279 | ||||||||||||||||||||
Literature Links: |
JMY PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
JMY - junction mediating and regulatory protein, p53 cofactor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152405.4 | 1281 | Missense Mutation | CAC,CAG | H,Q 244 | NP_689618.4 | |
XM_005248430.2 | 1281 | Missense Mutation | CAC,CAG | H,Q 244 | XP_005248487.1 | |
XM_011543155.2 | 1281 | Missense Mutation | CAC,CAG | H,Q 244 | XP_011541457.1 |