Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGAACCGGCTTGACTTTTTCATC[A/G]TCATCGCAGGGTGAGGACCTGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604065 | ||||||||||||||||||||
Literature Links: |
CACNA1G PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CACNA1G - calcium voltage-gated channel subunit alpha1 G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256324.1 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_001243253.1 | |
NM_001256325.1 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_001243254.1 | |
NM_001256326.1 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_001243255.1 | |
NM_001256327.1 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_001243256.1 | |
NM_001256328.1 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_001243257.1 | |
NM_001256329.1 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_001243258.1 | |
NM_001256330.1 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_001243259.1 | |
NM_001256331.1 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_001243260.1 | |
NM_001256332.1 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_001243261.1 | |
NM_001256333.1 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_001243262.1 | |
NM_001256334.1 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_001243263.1 | |
NM_001256359.1 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_001243288.1 | |
NM_001256360.1 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_001243289.1 | |
NM_001256361.1 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_001243290.1 | |
NM_018896.4 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_061496.2 | |
NM_198376.2 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_938190.1 | |
NM_198377.2 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_938191.2 | |
NM_198378.2 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_938192.1 | |
NM_198379.2 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_938193.1 | |
NM_198380.2 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_938194.1 | |
NM_198382.2 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_938196.1 | |
NM_198383.2 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_938197.1 | |
NM_198384.2 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_938198.1 | |
NM_198385.2 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_938199.1 | |
NM_198386.2 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_938200.1 | |
NM_198387.2 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_938201.1 | |
NM_198388.2 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_938202.1 | |
NM_198396.2 | 870 | Missense Mutation | ATC,GTC | I,V 160 | NP_938406.1 | |
XM_006722160.3 | 870 | Missense Mutation | ATC,GTC | I,V 160 | XP_006722223.1 | |
XM_006722161.3 | 870 | Missense Mutation | ATC,GTC | I,V 160 | XP_006722224.1 |
CACNA1G-AS1 - CACNA1G antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |