Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAATGTTTGGATGATAAATTGGA[A/G]TGAGAAATCGGATCTGAGGAGGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610538 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2T PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBE2T - ubiquitin conjugating enzyme E2 T | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310326.1 | 351 | Missense Mutation | ACT,ATT | T,I 42 | NP_001297255.1 | |
NM_014176.3 | 351 | Missense Mutation | ACT,ATT | T,I 72 | NP_054895.1 |