Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAAGTTCAGGACACGGGATTGAT[C/T]TATGACACTCCCACTTCGGAATCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602977 MIM: 191845 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GP2 - glycoprotein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UMOD - uromodulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008389.2 | 2209 | Missense Mutation | AAT,GAT | N,D 594 | NP_001008390.1 | |
NM_001278614.1 | 2209 | Missense Mutation | AAT,GAT | N,D 627 | NP_001265543.1 | |
NM_003361.3 | 2209 | Missense Mutation | AAT,GAT | N,D 594 | NP_003352.2 | |
XM_011545934.1 | 2209 | Missense Mutation | AAT,GAT | N,D 669 | XP_011544236.1 | |
XM_011545935.1 | 2209 | Missense Mutation | AAT,GAT | N,D 641 | XP_011544237.1 | |
XM_011545936.1 | 2209 | Missense Mutation | AAT,GAT | N,D 641 | XP_011544238.1 | |
XM_011545937.1 | 2209 | Missense Mutation | AAT,GAT | N,D 641 | XP_011544239.1 | |
XM_011545938.1 | 2209 | Missense Mutation | AAT,GAT | N,D 641 | XP_011544240.1 | |
XM_011545940.1 | 2209 | Missense Mutation | AAT,GAT | N,D 690 | XP_011544242.1 | |
XM_017023641.1 | 2209 | Missense Mutation | AAT,GAT | N,D 622 | XP_016879130.1 |