Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGGAAGAAAGAACCAAGCATA[A/C]ATGATTTAGTCCCCTCTCCTCTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CECR5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CECR5 - cat eye syndrome chromosome region, candidate 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017829.5 | 46 | Missense Mutation | ATG,ATT | M,I 1 | NP_060299.4 | |
NM_033070.2 | 46 | Intron | NP_149061.1 | |||
XM_005261236.3 | 46 | Intron | XP_005261293.1 | |||
XM_011546126.2 | 46 | Intron | XP_011544428.1 | |||
XM_011546127.1 | 46 | Intron | XP_011544429.1 | |||
XM_017028781.1 | 46 | Intron | XP_016884270.1 | |||
XM_017028782.1 | 46 | Intron | XP_016884271.1 | |||
XM_017028783.1 | 46 | Intron | XP_016884272.1 | |||
XM_017028784.1 | 46 | Intron | XP_016884273.1 |
CECR5-AS1 - CECR5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |