Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCAGGCCTGACACAATATCCCT[C/G]TTCACCTGCAGGTGGGAAGGAACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602843 MIM: 610642 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGDIB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGDIB - Rho GDP dissociation inhibitor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001175.6 | 461 | Missense Mutation | AAC,AAG | N,K 116 | NP_001166.3 | |
NM_001321420.1 | 461 | Missense Mutation | AAC,AAG | N,K 116 | NP_001308349.1 | |
NM_001321421.1 | 461 | Missense Mutation | AAC,AAG | N,K 116 | NP_001308350.1 | |
NM_001321422.1 | 461 | Missense Mutation | AAC,AAG | N,K 116 | NP_001308351.1 | |
NM_001321423.1 | 461 | Missense Mutation | AAC,AAG | N,K 116 | NP_001308352.1 |
ERP27 - endoplasmic reticulum protein 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |