Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAACAGATAAAACTGAAAGAGG[C/T]GGAGGGGACCCTCCTGCAGCCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602385 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SULT1C2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SULT1C2 - sulfotransferase family 1C member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001056.3 | 500 | Missense Mutation | GCG,GTG | A,V 16 | NP_001047.1 | |
NM_176825.2 | 500 | Missense Mutation | GCG,GTG | A,V 16 | NP_789795.1 | |
XM_005264013.3 | 500 | Missense Mutation | GCG,GTG | A,V 16 | XP_005264070.1 |