Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGGGATTAATGATCCCACTCTC[A/G]ATGTATTCGAAGACTGTGTATGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615365 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AK8 - adenylate kinase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317958.1 | 1334 | Silent Mutation | ATC,ATT | I,I 262 | NP_001304887.1 | |
NM_001317959.1 | 1334 | Silent Mutation | ATC,ATT | I,I 294 | NP_001304888.1 | |
NM_152572.2 | 1334 | Silent Mutation | ATC,ATT | I,I 466 | NP_689785.1 | |
XM_005272169.2 | 1334 | Silent Mutation | ATC,ATT | I,I 478 | XP_005272226.1 | |
XM_006716965.2 | 1334 | Silent Mutation | ATC,ATT | I,I 394 | XP_006717028.1 | |
XM_011518277.2 | 1334 | Intron | XP_011516579.1 | |||
XM_011518278.2 | 1334 | Silent Mutation | ATC,ATT | I,I 392 | XP_011516580.1 | |
XM_017014308.1 | 1334 | Silent Mutation | ATC,ATT | I,I 294 | XP_016869797.1 |