Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCTGGGAGCCCTGTCATGGGGC[A/G]GTGACTTCCATCCCGAAGCTGGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609011 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANGEL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANGEL1 - angel homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015305.3 | 2117 | Silent Mutation | ACC,ACT | T,T 668 | NP_056120.2 | |
XM_017021115.1 | 2117 | Intron | XP_016876604.1 |
LOC100506603 - uncharacterized LOC100506603 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VASH1 - vasohibin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |