Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGGTGGTATATACAGGAGGTGGA[G/A]GGACAGCAGGAAGCAAGGAGCCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616666 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHF12 - PHD finger protein 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEZ6 - seizure related 6 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098635.1 | 3165 | UTR 3 | NP_001092105.1 | |||
NM_001290202.1 | 3165 | UTR 3 | NP_001277131.1 | |||
NM_178860.4 | 3165 | UTR 3 | NP_849191.3 | |||
XM_011524315.1 | 3165 | Silent Mutation | CCC,CCT | P,P 985 | XP_011522617.1 | |
XM_011524317.2 | 3165 | UTR 3 | XP_011522619.1 |