Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAGTCGAGGGAGCGGCCGCTGCA[A/T]CCAGATGTGGTTTGACTCAGCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602469 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOLR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOLR3 - folate receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000804.3 | 822 | Missense Mutation | AAC,ATC | N,I 210 | NP_000795.2 | |
NM_001318045.1 | 822 | Missense Mutation | AAC,ATC | N,I 119 | NP_001304974.1 | |
XM_011544876.1 | 822 | Missense Mutation | AAC,ATC | N,I 118 | XP_011543178.1 |