Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGCTTCTTCAAACCAGAATATC[A/G]GGTTAGTATCTGGCAGGGAGGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601760 MIM: 612802 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2H4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTF2H4 - general transcription factor IIH subunit 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VARS2 - valyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167733.2 | 547 | UTR 5 | NP_001161205.1 | |||
NM_001167734.1 | 547 | Missense Mutation | CAG,CGG | Q,R 158 | NP_001161206.1 | |
NM_020442.5 | 547 | Missense Mutation | CAG,CGG | Q,R 128 | NP_065175.4 |