Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGAGACCACCACCGGGCTCCCAG[C/T]GAGGAAGAGGCCTTGGAGAAATGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615475 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHX34 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHX34 - DEAH-box helicase 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014681.5 | 358 | Silent Mutation | AGC,AGT | S,S 20 | NP_055496.2 | |
XM_005259443.3 | 358 | Silent Mutation | AGC,AGT | S,S 20 | XP_005259500.1 | |
XM_011527550.2 | 358 | Missense Mutation | AGC,AGT | S,S 20 | XP_011525852.1 | |
XM_011527551.2 | 358 | Missense Mutation | AGC,AGT | S,S 20 | XP_011525853.1 | |
XM_017027524.1 | 358 | UTR 5 | XP_016883013.1 |