Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGTCGGACATCGTGTCTCGCAC[C/T]GTGATGGTCCGGAAAGGACCCGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126375 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNMT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNMT1 - DNA (cytosine-5-)-methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130823.2 | 4382 | Silent Mutation | ACA,ACG | T,T 1382 | NP_001124295.1 | |
NM_001318730.1 | 4382 | Silent Mutation | ACA,ACG | T,T 1366 | NP_001305659.1 | |
NM_001318731.1 | 4382 | Silent Mutation | ACA,ACG | T,T 1261 | NP_001305660.1 | |
NM_001379.3 | 4382 | Silent Mutation | ACA,ACG | T,T 1366 | NP_001370.1 |