Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAAGGGATGATTGATGACACAGC[A/G]TTTCATCTTTATTACATTCACAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194538 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF10 - zinc finger protein 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF891 - zinc finger protein 891 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277291.1 | 1093 | Missense Mutation | ACG,ATG | T,M 301 | NP_001264220.1 | |
XM_017018666.1 | 1093 | Missense Mutation | ACG,ATG | T,M 301 | XP_016874155.1 | |
XM_017018667.1 | 1093 | Missense Mutation | ACG,ATG | T,M 301 | XP_016874156.1 | |
XM_017018668.1 | 1093 | Missense Mutation | ACG,ATG | T,M 301 | XP_016874157.1 |