Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTATTGTGGTCATAGTGACCTC[C/T]GACCCACACCTGCACACAACCATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR4D5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR4D5 - olfactory receptor family 4 subfamily D member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001965.1 | 153 | Silent Mutation | TCC,TCT | S,S 51 | NP_001001965.1 |
OR6T1 - olfactory receptor family 6 subfamily T member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |