Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCTACCCCGCCTGGTACCTCCG[C/T]TGCCATCCTCTCATCCAGGAACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606088 MIM: 605916 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JMJD7-PLA2G4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JMJD7-PLA2G4B - JMJD7-PLA2G4B readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLA2G4B - phospholipase A2 group IVB | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPTBN5 - spectrin beta, non-erythrocytic 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016642.3 | 10944 | Missense Mutation | ACG,GCG | T,A 3580 | NP_057726.4 | |
XM_017022299.1 | 10944 | Missense Mutation | ACG,GCG | T,A 3640 | XP_016877788.1 | |
XM_017022300.1 | 10944 | Missense Mutation | ACG,GCG | T,A 3600 | XP_016877789.1 | |
XM_017022301.1 | 10944 | Missense Mutation | ACG,GCG | T,A 3348 | XP_016877790.1 | |
XM_017022302.1 | 10944 | Missense Mutation | ACG,GCG | T,A 2699 | XP_016877791.1 | |
XM_017022303.1 | 10944 | Intron | XP_016877792.1 | |||
XM_017022304.1 | 10944 | Intron | XP_016877793.1 | |||
XM_017022305.1 | 10944 | Intron | XP_016877794.1 |