Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTAGAGCCCCAGCACAACAGTCA[A/C]GACCAGCATCACTGCAGCCATCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605702 MIM: 604673 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYVE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYVE1 - lymphatic vessel endothelial hyaluronan receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRVI1 - murine retrovirus integration site 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098579.2 | 2926 | Missense Mutation | GTG,TTG | V,L 857 | NP_001092049.2 | |
NM_001100163.2 | 2926 | Missense Mutation | GTG,TTG | V,L 774 | NP_001093633.1 | |
NM_001100167.2 | 2926 | Missense Mutation | GTG,TTG | V,L 550 | NP_001093637.1 | |
NM_001206880.1 | 2926 | Missense Mutation | GTG,TTG | V,L 659 | NP_001193809.1 | |
NM_001206881.1 | 2926 | Missense Mutation | GTG,TTG | V,L 550 | NP_001193810.1 | |
NM_130385.3 | 2926 | Missense Mutation | GTG,TTG | V,L 865 | NP_569056.4 |
MRVI1-AS1 - MRVI1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |