Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCGGAGGACCTGATGGGTGTGGC[A/G]GGGAGGGTGGGGGTGCCGGAGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601417 MIM: 611500 MIM: 602045 MIM: 601416 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD17B8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSD17B8 - hydroxysteroid 17-beta dehydrogenase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR219A1 - microRNA 219a-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RING1 - ring finger protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002931.3 | 1194 | Missense Mutation | AGG,GGG | R,G 334 | NP_002922.2 |
SLC39A7 - solute carrier family 39 member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |