Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCATTGAGGAAGCAGTAGAGGA[C/T]GGCCACCACCAGTCCCTGCCAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182098 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCTR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCTR - secretin receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002980.2 | 755 | Missense Mutation | ATC,GTC | I,V 386 | NP_002971.2 | |
XM_005263730.4 | 755 | Missense Mutation | ATC,GTC | I,V 208 | XP_005263787.1 | |
XM_011511621.2 | 755 | Missense Mutation | ATC,GTC | I,V 391 | XP_011509923.1 | |
XM_017004670.1 | 755 | Missense Mutation | ATC,GTC | I,V 381 | XP_016860159.1 | |
XM_017004671.1 | 755 | Missense Mutation | ATC,GTC | I,V 355 | XP_016860160.1 | |
XM_017004672.1 | 755 | Missense Mutation | ATC,GTC | I,V 323 | XP_016860161.1 | |
XM_017004673.1 | 755 | Missense Mutation | ATC,GTC | I,V 213 | XP_016860162.1 |
TMEM37 - transmembrane protein 37 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |