Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTGCGCCGCGCTGGCGGCCTTC[C/T]TGTTCGAGTACGACACGCCGCGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600846 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
P2RX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
P2RX4 - purinergic receptor P2X 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256796.1 | 338 | Silent Mutation | CTG,TTG | L,L 12 | NP_001243725.1 | |
NM_001261397.1 | 338 | Silent Mutation | CTG,TTG | L,L 12 | NP_001248326.1 | |
NM_001261398.1 | 338 | Silent Mutation | CTG,TTG | L,L 12 | NP_001248327.1 | |
NM_002560.2 | 338 | Silent Mutation | CTG,TTG | L,L 12 | NP_002551.2 | |
XM_011538416.2 | 338 | Silent Mutation | CTG,TTG | L,L 12 | XP_011536718.1 |