Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACACACGTCACTGGACCAAATAT[C/T]TCTTCCGTCATGCAGCAGGATTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606467 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH8A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH8A1 - aldehyde dehydrogenase 8 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193480.1 | 1092 | Silent Mutation | GAA,GAG | E,E 341 | NP_001180409.1 | |
NM_022568.3 | 1092 | Silent Mutation | GAA,GAG | E,E 391 | NP_072090.1 | |
NM_170771.2 | 1092 | Silent Mutation | GAA,GAG | E,E 337 | NP_739577.1 |