Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGAGTCCCTGACGTCCACCGGGC[A/G]GGTGCAGGCACCTAATTGGGCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605854 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BBC3 - BCL2 binding component 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127240.2 | 861 | Missense Mutation | CGC,TGC | R,C 233 | NP_001120712.1 | |
NM_001127241.2 | 861 | UTR 3 | NP_001120713.1 | |||
NM_001127242.2 | 861 | Missense Mutation | CGC,TGC | R,C 73 | NP_001120714.1 | |
NM_014417.4 | 861 | UTR 3 | NP_055232.1 | |||
XM_006723141.3 | 861 | UTR 3 | XP_006723204.1 | |||
XM_011526722.2 | 861 | UTR 3 | XP_011525024.1 |
MIR3190 - microRNA 3190 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3191 - microRNA 3191 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |