Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCTCTACACTGCCTATGTAGTC[A/G]TTGGTGCCATTGCTGGTGTTATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300545 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HS6ST2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HS6ST2 - heparan sulfate 6-O-sulfotransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077188.1 | 2321 | Silent Mutation | AAC,AAT | N,N 635 | NP_001070656.1 | |
NM_147175.3 | 2321 | Silent Mutation | AAC,AAT | N,N 595 | NP_671704.3 | |
XM_005262490.3 | 2321 | Silent Mutation | AAC,AAT | N,N 635 | XP_005262547.1 | |
XM_005262491.2 | 2321 | Silent Mutation | AAC,AAT | N,N 606 | XP_005262548.1 | |
XM_011531406.1 | 2321 | Silent Mutation | AAC,AAT | N,N 489 | XP_011529708.1 | |
XM_011531407.2 | 2321 | Intron | XP_011529709.1 | |||
XM_011531408.2 | 2321 | Intron | XP_011529710.1 | |||
XM_017029944.1 | 2321 | Silent Mutation | AAC,AAT | N,N 595 | XP_016885433.1 | |
XM_017029945.1 | 2321 | Silent Mutation | AAC,AAT | N,N 489 | XP_016885434.1 | |
XM_017029946.1 | 2321 | Silent Mutation | AAC,AAT | N,N 460 | XP_016885435.1 |