Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGAATTGTTAATTATAAAAACTC[A/G]GTATAGTAGTTCATAGTAATCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606648 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL22RA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL22RA2 - interleukin 22 receptor subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052962.2 | 815 | Nonsense Mutation | CGA,TGA | R,* 204 | NP_443194.1 | |
NM_181309.1 | 815 | Nonsense Mutation | CGA,TGA | R,* 172 | NP_851826.1 | |
NM_181310.1 | 815 | Intron | NP_851827.1 |