Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTTCGACCCCTGCATGTACCGC[C/G]AGTTGCTGCTAGTGCACCGCTTCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615315 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B3GNT6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B3GNT6 - UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138706.4 | 1189 | Missense Mutation | CAG,GAG | Q,E 351 | NP_619651.3 |