Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGCGCGCCTGAGGCCTCCTTGA[C/T]CGGGCCGTCGGGCGCGGCGGGGACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606199 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTD-2194D22.4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTD-2194D22.4 - uncharacterized LOC101929081 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IRX4 - iroquois homeobox 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278632.1 | 1107 | Missense Mutation | ATC,GTC | I,V 297 | NP_001265561.1 | |
NM_001278633.1 | 1107 | Missense Mutation | ATC,GTC | I,V 323 | NP_001265562.1 | |
NM_001278634.1 | 1107 | Missense Mutation | ATC,GTC | I,V 297 | NP_001265563.1 | |
NM_001278635.1 | 1107 | Missense Mutation | ATC,GTC | I,V 323 | NP_001265564.1 | |
NM_016358.2 | 1107 | Missense Mutation | ATC,GTC | I,V 297 | NP_057442.1 |