Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAGAAGCTGATGTAATTACCTTT[C/T]AAGAAACATCATGAAGTATAACAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608989 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDR16C5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SDR16C5 - short chain dehydrogenase/reductase family 16C, member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318049.1 | 1564 | Missense Mutation | AAA,GAA | K,E 278 | NP_001304978.1 | |
NM_001318050.1 | 1564 | Missense Mutation | AAA,GAA | K,E 234 | NP_001304979.1 | |
NM_138969.3 | 1564 | Missense Mutation | AAA,GAA | K,E 278 | NP_620419.2 | |
XM_011517479.1 | 1564 | Missense Mutation | AAA,GAA | K,E 234 | XP_011515781.1 |