Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTAAACAGTCCAACACCTCTTCT[G/T]TTAGCAAGCCTAGGCAAGGGGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
IZUMO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IZUMO2 - IZUMO family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321449.1 | 512 | Missense Mutation | AAA,ACA | K,T 142 | NP_001308378.1 | |
NM_152358.2 | 512 | Missense Mutation | AAA,ACA | K,T 142 | NP_689571.2 | |
XM_011526436.2 | 512 | Missense Mutation | AAA,ACA | K,T 142 | XP_011524738.1 | |
XM_011526437.2 | 512 | Missense Mutation | AAA,ACA | K,T 135 | XP_011524739.1 |