Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTCCTTAGGTTGATATGGAGGT[A/G]ACTCTTCCAGGCGAGGGTAAAGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607356 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGO4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGO4 - argonaute 4, RISC catalytic component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017629.3 | 396 | Silent Mutation | GTA,GTG | V,V 107 | NP_060099.2 | |
XM_005270578.3 | 396 | Silent Mutation | GTA,GTG | V,V 40 | XP_005270635.1 | |
XM_005270579.3 | 396 | Silent Mutation | GTA,GTG | V,V 40 | XP_005270636.1 | |
XM_011540883.2 | 396 | Intron | XP_011539185.1 | |||
XM_011540884.2 | 396 | Intron | XP_011539186.1 | |||
XM_017000529.1 | 396 | Silent Mutation | GTA,GTG | V,V 107 | XP_016856018.1 |