Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATCCGAAAGGCGTACCTGGAAGA[C/T]TTTTACACCCGGATGATGCATGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616713 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIPOX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIPOX - pipecolic acid and sarcosine oxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016518.2 | 512 | Silent Mutation | GAC,GAT | D,D 62 | NP_057602.2 | |
XM_017024722.1 | 512 | Intron | XP_016880211.1 |