Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTGCTGAAAACTCCAGTCTTAT[G/T]AAACACTTGAGAGTTCACACAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF586 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF586 - zinc finger protein 586 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077426.2 | 941 | Nonsense Mutation | TGA,TTA | *,L 208 | NP_001070894.1 | |
NM_001204814.1 | 941 | Missense Mutation | ATG,ATT | M,I 207 | NP_001191743.1 | |
NM_017652.3 | 941 | Missense Mutation | ATG,ATT | M,I 250 | NP_060122.2 |