Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCAGAAGATCATGGGACTTGTG[A/G]ACCAGGACCTGGAGCAGCACGTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608411 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
XPO6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XPO6 - exportin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270940.1 | 3138 | Silent Mutation | GTC,GTT | V,V 1021 | NP_001257869.1 | |
NM_015171.3 | 3138 | Silent Mutation | GTC,GTT | V,V 1035 | NP_055986.1 | |
XM_005255197.3 | 3138 | Silent Mutation | GTC,GTT | V,V 961 | XP_005255254.1 | |
XM_005255198.1 | 3138 | Silent Mutation | GTC,GTT | V,V 861 | XP_005255255.1 | |
XM_005255199.2 | 3138 | Silent Mutation | GTC,GTT | V,V 690 | XP_005255256.1 | |
XM_011545772.2 | 3138 | Intron | XP_011544074.1 |