Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGAGGAGACCGCCGCTGCCACC[C/G]TGCTACTGCAGGGTGAGGAAGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600473 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01024 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01024 - long intergenic non-protein coding RNA 1024 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PURA - purine rich element binding protein A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005859.4 | 990 | Missense Mutation | CTG,GTG | L,V 311 | NP_005850.1 |