Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCTGACTTGATGGTGAATCTGCC[A/G]AAGAGGGCAGCTTCTGGGGCTGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615790 MIM: 164940 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AHDC1 - AT-hook DNA binding motif containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FGR - FGR proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042729.1 | 1524 | Silent Mutation | TTC,TTT | F,F 432 | NP_001036194.1 | |
NM_001042747.1 | 1524 | Silent Mutation | TTC,TTT | F,F 432 | NP_001036212.1 | |
NM_005248.2 | 1524 | Silent Mutation | TTC,TTT | F,F 432 | NP_005239.1 | |
XM_006710452.2 | 1524 | Silent Mutation | TTC,TTT | F,F 432 | XP_006710515.1 | |
XM_011541010.1 | 1524 | Silent Mutation | TTC,TTT | F,F 432 | XP_011539312.1 | |
XM_011541011.2 | 1524 | Silent Mutation | TTC,TTT | F,F 363 | XP_011539313.1 | |
XM_011541012.1 | 1524 | Missense Mutation | TCG,TTG | S,L 381 | XP_011539314.1 | |
XM_011541013.2 | 1524 | UTR 3 | XP_011539315.1 | |||
XM_011541014.2 | 1524 | Silent Mutation | TTC,TTT | F,F 228 | XP_011539316.1 | |
XM_017000673.1 | 1524 | Silent Mutation | TTC,TTT | F,F 228 | XP_016856162.1 | |
XM_017000674.1 | 1524 | Silent Mutation | TTC,TTT | F,F 228 | XP_016856163.1 |