Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCAGAAAGGGAGGAATCGGAGCC[A/C]CCTTTTGTGGACCTGAGAGAAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608441 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYNE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYNE1 - spectrin repeat containing nuclear envelope protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033071.3 | 26626 | Silent Mutation | GGG,GGT | G,G 8674 | NP_149062.1 | |
NM_182961.3 | 26626 | Silent Mutation | GGG,GGT | G,G 8722 | NP_892006.3 | |
XM_006715407.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8771 | XP_006715470.1 | |
XM_006715408.2 | 26626 | Silent Mutation | GGG,GGT | G,G 8767 | XP_006715471.1 | |
XM_006715409.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8764 | XP_006715472.1 | |
XM_006715410.2 | 26626 | Silent Mutation | GGG,GGT | G,G 8757 | XP_006715473.1 | |
XM_006715411.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8754 | XP_006715474.1 | |
XM_006715412.2 | 26626 | Silent Mutation | GGG,GGT | G,G 8752 | XP_006715475.1 | |
XM_006715413.2 | 26626 | Silent Mutation | GGG,GGT | G,G 8748 | XP_006715476.1 | |
XM_006715414.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8747 | XP_006715477.1 | |
XM_006715415.2 | 26626 | Silent Mutation | GGG,GGT | G,G 8734 | XP_006715478.1 | |
XM_006715416.2 | 26626 | Silent Mutation | GGG,GGT | G,G 8729 | XP_006715479.1 | |
XM_006715417.2 | 26626 | Silent Mutation | GGG,GGT | G,G 8724 | XP_006715480.1 | |
XM_006715420.2 | 26626 | Silent Mutation | GGG,GGT | G,G 8720 | XP_006715483.1 | |
XM_006715421.2 | 26626 | Silent Mutation | GGG,GGT | G,G 8719 | XP_006715484.1 | |
XM_006715422.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8718 | XP_006715485.1 | |
XM_006715423.2 | 26626 | Missense Mutation | GGG,TGG | G,W 8752 | XP_006715486.1 | |
XM_006715424.2 | 26626 | Missense Mutation | GGG,TGG | G,W 8738 | XP_006715487.1 | |
XM_006715425.2 | 26626 | Missense Mutation | GGG,TGG | G,W 8715 | XP_006715488.1 | |
XM_011535641.2 | 26626 | Silent Mutation | GGG,GGT | G,G 8770 | XP_011533943.1 | |
XM_011535642.2 | 26626 | Silent Mutation | GGG,GGT | G,G 8766 | XP_011533944.1 | |
XM_011535643.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8716 | XP_011533945.1 | |
XM_011535644.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8196 | XP_011533946.1 | |
XM_011535645.2 | 26626 | Silent Mutation | GGG,GGT | G,G 8027 | XP_011533947.1 | |
XM_017010608.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8771 | XP_016866097.1 | |
XM_017010609.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8771 | XP_016866098.1 | |
XM_017010610.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8764 | XP_016866099.1 | |
XM_017010611.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8762 | XP_016866100.1 | |
XM_017010612.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8745 | XP_016866101.1 | |
XM_017010613.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8733 | XP_016866102.1 | |
XM_017010614.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8719 | XP_016866103.1 | |
XM_017010615.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8682 | XP_016866104.1 | |
XM_017010616.1 | 26626 | Missense Mutation | GGG,TGG | G,W 8729 | XP_016866105.1 | |
XM_017010617.1 | 26626 | Missense Mutation | GGG,TGG | G,W 8714 | XP_016866106.1 | |
XM_017010618.1 | 26626 | Missense Mutation | GGG,TGG | G,W 8710 | XP_016866107.1 | |
XM_017010619.1 | 26626 | Silent Mutation | GGG,GGT | G,G 8196 | XP_016866108.1 |